Skip to Content
Merck
All Photos(1)

Key Documents

8.42712

Sigma-Aldrich

3,6-Dichloropyridazine

for synthesis

Synonym(s):

3,6-Dichloropyridazine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 74.40
100 G
CHF 175.00

CHF 74.40


0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
CHF 74.40
100 G
CHF 175.00

About This Item

Empirical Formula (Hill Notation):
C4H2Cl2N2
CAS Number:
Molecular Weight:
148.98
MDL number:
UNSPSC Code:
12352101
EC Index Number:
205-478-6
NACRES:
NA.22

CHF 74.40


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

solid

mp

72 °C

storage temp.

2-30°C

SMILES string

Clc1nnc(cc1)Cl

InChI

1S/C4H2Cl2N2/c5-3-1-2-4(6)8-7-3/h1-2H

InChI key

GUSWJGOYDXFJSI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 66 °C
Melting range (upper value): ≤ 69 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service