Skip to Content
Merck
All Photos(1)

Key Documents

8.42593

Sigma-Aldrich

2-Nitrobenzenesulfonyl chloride

for synthesis

Synonym(s):

2-Nitrobenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 54.20
100 G
CHF 119.00

CHF 54.20


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
CHF 54.20
100 G
CHF 119.00

About This Item

Empirical Formula (Hill Notation):
C6H4ClNO4S
CAS Number:
Molecular Weight:
221.62
MDL number:
UNSPSC Code:
12352005
EC Index Number:
216-907-1
NACRES:
NA.22

CHF 54.20


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

solid

mp

65-67 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(Cl)c1c(cccc1)[N+](=O)[O-]

InChI

1S/C6H4ClNO4S/c7-13(11,12)6-4-2-1-3-5(6)8(9)10/h1-4H

InChI key

WPHUUIODWRNJLO-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Melting range (lower value): ≥ 65 °C
Melting range (upper value): ≤ 68 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service