Skip to Content
Merck
All Photos(1)

Key Documents

8.41566

Sigma-Aldrich

Benzyl phenyl sulfide

for synthesis

Synonym(s):

Benzyl phenyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 148.00

CHF 148.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
CHF 148.00

About This Item

Empirical Formula (Hill Notation):
C13H12S
CAS Number:
Molecular Weight:
200.30
MDL number:
UNSPSC Code:
12352113
EC Index Number:
212-612-7
NACRES:
NA.22

CHF 148.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

powder

mp

40-44 °C

storage temp.

2-30°C

SMILES string

S(Cc2ccccc2)c1ccccc1

InChI

1S/C13H12S/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

LKMCJXXOBRCATQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 41 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service