Skip to Content
Merck
All Photos(1)

Key Documents

8.41513

Sigma-Aldrich

5α-Cholestan-3ß-ol

for synthesis

Synonym(s):

5α-Cholestan-3ß-ol, Dihydrocholesterol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 110.00

CHF 110.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 G
CHF 110.00

About This Item

Empirical Formula (Hill Notation):
C27H48O
CAS Number:
Molecular Weight:
388.67
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-315-8
NACRES:
NA.22

CHF 110.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥97.5% (GC)

form

solid

mp

130-143 °C

storage temp.

2-30°C

SMILES string

OC1CC2C(C3C(C4C(C(CC4)C(CCCC(C)C)C)(CC3)C)CC2)(CC1)C

InChI

1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3

InChI key

QYIXCDOBOSTCEI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Melting range (lower value): ≥ 139 °C
Melting range (upper value): ≤ 143 °C
Spec. rotation [α²0/D (c=1 in chloroform): +22.0 - +25.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service