840651
Probumin® Bovine Serum Albumin Vaccine Grade
≥95%, USA origin, moisture (loss on drying): 0-10% ; heavy metals : 0-50 μg/g
Synonym(s):
Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V
About This Item
Recommended Products
product name
Probumin® Bovine Serum Albumin Vaccine Grade, package of 10 g
biological source
bovine serum
Quality Level
Assay
≥95% (purity)
form
lyophilized powder
shelf life
48 mo.
mol wt
~66 kDa
purified by
heat shock fractionation
packaging
package of 10 g
manufacturer/tradename
Probumin®
origin
USA origin
technique(s)
microbiological culture: suitable
dilution
(Adjuvant for veterinary use)
impurities
0-10% moisture
0-50 μg/g heavy metals
loss
≤5%
pH
5-6
solubility
water: soluble 40 mg/mL
cation traces
0-50 μg/mg
foreign activity
Tested for E. coli
Tested for Mycoplasma
Tested for Salmonella
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Disclaimer
For further manufacturing use only. Not for human or animal consumption.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service