Skip to Content
Merck
All Photos(1)

Key Documents

8.40160

Sigma-Aldrich

2-Isopropylphenol

for synthesis

Synonym(s):

2-Isopropylphenol, 2-(1-Methylethyl)phenol, 2-Hydroxycumene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 65.40

CHF 65.40


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
CHF 65.40

About This Item

Empirical Formula (Hill Notation):
C9H12O
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-852-8
NACRES:
NA.22

CHF 65.40


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

<0.066 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

212-214 °C/1013 hPa

mp

14-16 °C

transition temp

flash point 88 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(cccc1)C(C)C

InChI

1S/C9H12O/c1-7(2)8-5-3-4-6-9(8)10/h3-7,10H,1-2H3

InChI key

CRBJBYGJVIBWIY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.993 - 0.997
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service