Skip to Content
Merck
All Photos(1)

Key Documents

8310-OP

Millipore

SSC Buffer

OmniPur® Grade, 20X Liquid Concentrate

Synonym(s):

Saline-Sodium Citrate Buffer, 20X

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
inquire


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4 L
inquire

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

storage temp.

10-30°C

General description



General Composition: Each liter of concentrate makes 20 L of 1X SSC buffer (1X = 0.150 M Sodium Chloride, 0.015 M Sodium Citrate). Molecular Biology grade.

DNase: None detected
pH of solution at 25°C: 6.85-7.15
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service