Skip to Content
Merck
All Photos(2)

Key Documents

8.21967

Sigma-Aldrich

4′-Bromoacetanilide

for synthesis

Synonym(s):

4′-Bromoacetanilide, N-Acetyl-p-bromoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(Br)C6H4NHCOCH3
CAS Number:
Molecular Weight:
214.06
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-154-9
form:
powder

Quality Level

form

powder

mp

164-167 °C

storage temp.

2-30°C

InChI

1S/C8H8BrNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)

InChI key

MSLICLMCQYQNPK-UHFFFAOYSA-N

Analysis Note

Assay (ex Br): ≥ 98.0 %
Melting range (lower value): ≥ 164 °C
Melting range (upper value): ≤ 167 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service