Skip to Content
Merck
All Photos(1)

Key Documents

8.21935

Sigma-Aldrich

1-Ethylquinaldinium iodide

for synthesis

Synonym(s):

1-Ethylquinaldinium iodide, N-Ethyl-2-methylquinolinium iodide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
CHF 121.00

CHF 121.00


Estimated to ship on24 March 2025


Request a Bulk Order

Select a Size

Change View
50 G
CHF 121.00

About This Item

Empirical Formula (Hill Notation):
C12H14IN
CAS Number:
Molecular Weight:
299.15
MDL number:
UNSPSC Code:
12352107
EC Index Number:
210-120-7
NACRES:
NA.22

CHF 121.00


Estimated to ship on24 March 2025


Request a Bulk Order

Quality Level

form

powder

solubility

250 g/L

storage temp.

2-30°C

SMILES string

[I-].[n+]1(c2c(ccc1C)cccc2)CC

InChI

1S/C12H14N.HI/c1-3-13-10(2)8-9-11-6-4-5-7-12(11)13;/h4-9H,3H2,1-2H3;1H/q+1;/p-1

InChI key

OEVSHJVOKFWBJY-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service