Skip to Content
Merck
All Photos(1)

Key Documents

8.21095

Sigma-Aldrich

Tetrahydrothiophene

for synthesis

Synonym(s):

Tetrahydrothiophene, Tetramethylene sulfide, THT

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CHF 20.30
250 ML
CHF 50.50

CHF 20.30


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 ML
CHF 20.30
250 ML
CHF 50.50

About This Item

Empirical Formula (Hill Notation):
C4H8S
CAS Number:
Molecular Weight:
88.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-728-9
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

CHF 20.30


Only 0 left in stock

Request a Bulk Order

vapor pressure

19 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

202 °C

potency

1850 mg/kg LD50, oral (Rat)

expl. lim.

1.1-12.1 % (v/v)

pH

7 (20 °C in H2O)

bp

121 °C/1013 hPa

mp

-96 °C

transition temp

flash point 19 °C

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S1CCCC1

InChI

1S/C4H8S/c1-2-4-5-3-1/h1-4H2

InChI key

RAOIDOHSFRTOEL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.997 - 0.999
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service