Skip to Content
Merck
All Photos(1)

Key Documents

8.20953

Sigma-Aldrich

n-Pentadecane

for synthesis

Synonym(s):

n-Pentadecane

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
CHF 33.70
50 ML
CHF 62.10

CHF 33.70


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
CHF 33.70
50 ML
CHF 62.10

About This Item

Linear Formula:
CH3(CH2)13CH3
CAS Number:
Molecular Weight:
212.41
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-098-1
NACRES:
NA.22

CHF 33.70


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

0.000001 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

expl. lim.

0.45-6.5 % (v/v)

kinematic viscosity

<7 cSt(40 °C)

bp

270 °C/1013 hPa

mp

10 °C

transition temp

flash point 132.2 °C

density

0.77 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCC)CCCCCCC

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3

InChI key

YCOZIPAWZNQLMR-UHFFFAOYSA-N

Application

  • Densities and refractive indices of 1-hexanol+ n-pentadecane binary system at temperatures from (293.15 to 323.15) K: Examines the physical properties of n-Pentadecane mixtures, providing data useful for formulating binary solvent systems in chemical research (Crisciu et al., 2014).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.768 - 0.769
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service