Skip to Content
Merck
All Photos(1)

Key Documents

8.20617

Sigma-Aldrich

O-Benzylhydroxylamine

for synthesis

Synonym(s):

O-Benzylhydroxylamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
CHF 95.00

CHF 95.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
CHF 95.00

About This Item

Empirical Formula (Hill Notation):
C7H9NO
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

CHF 95.00


Estimated to ship on31 May 2025


Request a Bulk Order

vapor pressure

2.09 hPa ( 58 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

260 °C (DIN 51794)

pH

8 (47 g/L in H2O)

kinematic viscosity

3.34 cSt(23 °C)

bp

49 °C/1 hPa

transition temp

flash point 91.5 °C

density

1.064 g/cm3 at 23 °C

storage temp.

2-30°C

SMILES string

Cl.NOCc1ccccc1

InChI

1S/C7H9NO.ClH/c8-9-6-7-4-2-1-3-5-7;/h1-5H,6,8H2;1H

InChI key

HYDZPXNVHXJHBG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.062 - 1.066
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this compound soluble in water? or any common organic solvents?

    1 answer
    1. This product is soluble at 47 g/L in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service