Skip to Content
Merck
All Photos(1)

Documents

8.20616

Sigma-Aldrich

n-Heptadecane

for synthesis

Synonym(s):

n-Heptadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH3
CAS Number:
Molecular Weight:
240.47
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-108-4
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

kinematic viscosity

<7 cSt(40 °C)

bp

302 °C/1013 hPa

mp

22 °C

transition temp

flash point 148 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C17H36/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3-17H2,1-2H3

InChI key

NDJKXXJCMXVBJW-UHFFFAOYSA-N

Application


  • Zonisamide for Parkinson′s Disease Dyskinesia: Research on Parkinson′s disease patients with wearing-off symptoms identifies risk factors for dyskinesia when treated with Zonisamide, supporting its potential application in managing motor complications associated with the disease (Mishima et al., 2023).

  • Zonisamide Property Calculations: Research provides calculated descriptors for Zonisamide among other compounds, aiding in the development of better pharmacological profiles and enhancing its utility in pharmaceutical research for antiepileptic mechanisms (Liu, Acree, and Abraham, 2022).

  • Zonisamide Quantification in Therapy: A method using LC-MS/MS for the quantification of Zonisamide from dried sample spots demonstrates advancements in therapeutic drug monitoring, ensuring effective dosing and patient safety in clinical settings (DʼUrso et al., 2019).



Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service