Skip to Content
Merck
All Photos(1)

Key Documents

8.18819

Sigma-Aldrich

Triethoxymethylsilane

for synthesis

Synonym(s):

Triethoxymethylsilane, Methyltriethoxysilane, MTES

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
CHF 70.40

CHF 70.40


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
CHF 70.40

About This Item

Linear Formula:
(C2H5O)3SiCH3
CAS Number:
Molecular Weight:
178.30
MDL number:
UNSPSC Code:
12352103
EC Index Number:
217-983-9
NACRES:
NA.22

CHF 70.40


Estimated to ship on23 May 2025


Request a Bulk Order

vapor pressure

13.3 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

220 °C

potency

15700 mg/kg LD50, oral (Rat)

bp

142 °C/1013 hPa

mp

<-40 °C

transition temp

flash point 33 °C

density

0.89 g/cm3 at 20 °C

SMILES string

[Si](OCC)(OCC)(OCC)C

InChI

1S/C7H18O3Si/c1-5-8-11(4,9-6-2)10-7-3/h5-7H2,1-4H3

InChI key

CPUDPFPXCZDNGI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.894 - 0.896
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service