Skip to Content
Merck
All Photos(1)

Documents

8.18522

Sigma-Aldrich

3-Amino-1,2-propanediol

for synthesis

Synonym(s):

3-Amino-1,2-propanediol, 1-Amino-2,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
91.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
210-475-8
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

7500 mg/kg LD50, oral (Rat)

pH

11 (20 °C, 100 g/L in H2O)

transition temp

flash point 138 °C

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H9NO2/c4-1-3(6)2-5/h3,5-6H,1-2,4H2

InChI key

KQIGMPWTAHJUMN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.185 - 1.187
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service