Skip to Content
Merck
All Photos(1)

Key Documents

8.18107

Sigma-Aldrich

4,4-Dimethyl-2-oxazoline

for synthesis

Synonym(s):

4,4-Dimethyl-2-oxazoline

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CHF 156.00

CHF 156.00


Estimated to ship on06 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
CHF 156.00

About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
250-045-7
NACRES:
NA.22

CHF 156.00


Estimated to ship on06 May 2025


Request a Bulk Order

Quality Level

Assay

≥93.0% (GC)

form

liquid

bp

99-100 °C/1013 hPa

transition temp

flash point 1 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1=COCC1(C)C

InChI

1S/C5H9NO/c1-5(2)3-7-4-6-5/h4H,3H2,1-2H3

InChI key

KOAMXHRRVFDWRQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 93.0 % (a/a)
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F

Flash Point(C)

1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service