Skip to Content
Merck
All Photos(1)

Key Documents

8.18054

Sigma-Aldrich

Triphenylphosphine selenide

for synthesis

Synonym(s):

Triphenylphosphine selenide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
CHF 119.00

CHF 119.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 G
CHF 119.00

About This Item

Linear Formula:
(C6H5)3PSe
CAS Number:
Molecular Weight:
341.25
MDL number:
UNSPSC Code:
12352103
EC Index Number:
223-406-1
NACRES:
NA.22

CHF 119.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

crystals

mp

186-189 °C

storage temp.

2-30°C

SMILES string

[Se-][P+](c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C18H15PSe/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

ZFVJLNKVUKIPPI-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 99
Identity (IR-spectrum): passes test
Melting range: 186 - 189 °C

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service