Skip to Content
Merck
All Photos(1)

Key Documents

8.16100

Sigma-Aldrich

5-Chloro-2-(2,4-dichlorophenoxy)phenol

for synthesis

Synonym(s):

5-Chloro-2-(2,4-dichlorophenoxy)phenol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 159.00

CHF 159.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
CHF 159.00

About This Item

Empirical Formula (Hill Notation):
C12H7Cl3O2
CAS Number:
Molecular Weight:
289.54
MDL number:
UNSPSC Code:
12352104
EC Index Number:
222-182-2
NACRES:
NA.22

CHF 159.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

powder

potency

3700 mg/kg LD50, oral (Rat)
9300 mg/kg LD50, skin (Rabbit)

mp

55-60 °C

solubility

0.01 g/L

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)Oc2c(cc(cc2)Cl)O

InChI

1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H

InChI key

XEFQLINVKFYRCS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 55 °C
Melting range (upper value): ≤ 58 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service