Skip to Content
Merck
All Photos(2)

Key Documents

8.14961

Sigma-Aldrich

4′-Hydroxybiphenyl-4-carbonitrile

for synthesis

Synonym(s):

4′-Hydroxybiphenyl-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352117
EC Index Number:
427-840-4
NACRES:
NA.22

vapor pressure

0.022 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

mp

193-199 °C

solubility

12.7 g/L

storage temp.

2-30°C

SMILES string

N#Cc1ccc(cc1)c2ccc(cc2)O

InChI

1S/C13H9NO/c14-9-10-1-3-11(4-2-10)12-5-7-13(15)8-6-12/h1-8,15H

InChI key

ZRMIETZFPZGBEB-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 195 °C
Melting range (upper value): ≤ 199 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service