Skip to Content
Merck
All Photos(1)

Key Documents

8.14924

Sigma-Aldrich

1,2,3,4-Butanetetracarboxylic acid

for synthesis

Synonym(s):

1,2,3,4-Butanetetracarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
CHF 43.90
250 G
CHF 136.00

CHF 43.90


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
50 G
CHF 43.90
250 G
CHF 136.00

About This Item

Empirical Formula (Hill Notation):
C8H10O8
CAS Number:
Molecular Weight:
234.16
MDL number:
UNSPSC Code:
12352106
EC Index Number:
216-938-0
NACRES:
NA.22

CHF 43.90


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

potency

1720 mg/kg LD50, oral (Rat)

mp

187-190 °C

solubility

118 g/L

storage temp.

2-30°C

SMILES string

OC(=O)C(C(CC(=O)O)C(=O)O)CC(=O)O

InChI

1S/C8H10O8/c9-5(10)1-3(7(13)14)4(8(15)16)2-6(11)12/h3-4H,1-2H2,(H,9,10)(H,11,12)(H,13,14)(H,15,16)

InChI key

GGAUUQHSCNMCAU-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 186 °C
Melting range (upper value): ≤ 190 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service