Skip to Content
Merck
All Photos(1)

Documents

8.14868

Sigma-Aldrich

Trioctylphosphinoxide

for synthesis

Synonym(s):

Trioctylphosphinoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H17)3PO
CAS Number:
Molecular Weight:
386.63
MDL number:
UNSPSC Code:
12352119
EC Index Number:
201-121-3

Assay

≥98.0% (GC)

form

crystals

autoignition temp.

270 °C

potency

>10000 mg/kg LD50, oral (Rat)
2830 mg/kg LD50, skin (Rat)

bp

212 °C/39.99 Pa

mp

50-54 °C

transition temp

flash point 252 °C

density

0.88 g/cm3 at 25 °C

storage temp.

2-30°C

InChI

1S/C24H51OP/c1-4-7-10-13-16-19-22-26(25,23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

ZMBHCYHQLYEYDV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 50 °C
Melting range (upper value): ≤ 55 °C
Water (K. F.): ≤ 0.50 %
Acid value: ≤ 3
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

230 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service