Skip to Content
Merck
All Photos(1)

Key Documents

8.14553

Sigma-Aldrich

2-(Diisopropylamino)-ethylchloride hydrochloride

for synthesis

Synonym(s):

2-(Diisopropylamino)-ethylchloride hydrochloride, N-(2-Chloroethyl)-diisopropylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 71.10
250 G
CHF 141.00

CHF 71.10


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
CHF 71.10
250 G
CHF 141.00

About This Item

Empirical Formula (Hill Notation):
C8H19Cl2N
CAS Number:
Molecular Weight:
200.15
MDL number:
UNSPSC Code:
12352107
EC Index Number:
224-238-1
NACRES:
NA.22

CHF 71.10


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

form

crystals

potency

96.75 mg/kg LD50, oral (Rat)
197 mg/kg LD50, skin (Rabbit)

pH

4.0-4.5 (20 °C, 10 g/L in H2O)

mp

131-134 °C

storage temp.

2-30°C

SMILES string

[Cl-].ClCCN(C(C)C)C(C)C.[H+]

InChI

1S/C8H18ClN.ClH/c1-7(2)10(6-5-9)8(3)4;/h7-8H,5-6H2,1-4H3;1H

InChI key

IUSXYVRFJVAVOB-UHFFFAOYSA-N

General description

2-(Diisopropylamino)-ethylchloride hydrochloride commonly used as a building block in alkylation reactions to introduce diisopropylaminoethyl groups into various substrates, it can act as a ligand in the formation of metal complexes.

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 130 °C
Melting range (upper value): ≤ 133 °C
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service