Skip to Content
Merck
All Photos(1)

Key Documents

8.14490

Sigma-Aldrich

2,2-Dichlorobutyrophenone

for synthesis

Synonym(s):

2,2-Dichlorobutyrophenone, 2,2-Dichloropropyl phenyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CHF 123.00

CHF 123.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 ML
CHF 123.00

About This Item

CAS Number:
Molecular Weight:
217.09
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.22

CHF 123.00


Estimated to ship on29 May 2025


Quality Level

form

liquid

bp

132-136 °C

density

1.23 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(Cl)(CC)C(=O)c1ccccc1

InChI

1S/C10H10Cl2O/c1-2-10(11,12)9(13)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

VGWVNIXLMXHWCR-UHFFFAOYSA-N

General description

for synthesis

Application

2,2-Dichlorobutyrophenone for synthesis. CAS 66255-85-4, chemical formula C₆H₅COC(Cl₂)CH₂CH₃.

Analysis Note

Assay (GC, area%)≥ 96.0 % (a/a)Density (d 20 °C/ 4 °C)1.230 - 1.232Identity (IR)passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service