Skip to Content
Merck
All Photos(1)

Key Documents

8.14172

Sigma-Aldrich

Polyethylene glycol dimethyl ether 2000

for synthesis

Synonym(s):

Polyethylene glycol dimethyl ether 2000

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CHF 161.00

CHF 161.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
CHF 161.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

CHF 161.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

385 °C (DIN 51794)

kinematic viscosity

30 cSt(100 °C)

bp

>250 °C/1013 hPa

mp

49-53 °C

transition temp

flash point 254 °C

density

1.08 g/cm3 at 60 °C

storage temp.

2-30°C

SMILES string

O(CCOC)C

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Analysis Note

Hydroxyl value: ≤ 3
Melting range (lower value): ≥ 49 °C
Melting range (upper value): ≤ 54 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service