Skip to Content
Merck
All Photos(1)

Documents

8.05852

Sigma-Aldrich

1-Methylimidazole

for synthesis

Synonym(s):

1-Methylimidazole, N-Methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
MDL number:
UNSPSC Code:
12352005
EC Index Number:
210-484-7
NACRES:
NA.22

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

525 °C

potency

1144 mg/kg LD50, oral (Rat)
400-640 mg/kg LD50, skin (Rabbit)

expl. lim.

2.7-15.7 % (v/v)

pH

9.5-10.5 (20 °C, 50 g/L in H2O)

bp

198 °C/1013 hPa

mp

-2 °C

transition temp

flash point 92 °C

density

1.035 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Application

  • Enhancement of noncorrosive electrolytes: Research discusses the role of 1-Methylimidazole in improving the properties of noncorrosive electrolytes for battery applications. This study explores how 1-Methylimidazole acts as a high donor number cosolvent, aiding in the release of free anions from commercially available magnesium salts, which could lead to innovations in battery technology (Xiao et al., 2024).
  • Catalytic applications in organic synthesis: 1-Methylimidazole is utilized as a ligand in an air-stable ruthenium(II) complex, facilitating catalytic transfer hydrogenation of ketones from ethanol. This application demonstrates the catalytic versatility of 1-Methylimidazole in synthetic organic chemistry, providing a method for more efficient and sustainable chemical reactions (Beaufils et al., 2024).
  • Functional materials for biomedical applications: Research involving 1-Methylimidazole focuses on developing sodium alginate- and cationic cellulose-functionalized polycaprolactone nanofibers. These materials are designed for specific in vitro and antibacterial applications, indicating the utility of 1-Methylimidazole in the creation of functional biomedical materials (Tolba et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.034 - 1.036
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service