Skip to Content
Merck
All Photos(1)

Documents

8.05796

Sigma-Aldrich

3-Methoxybenzaldehyde

for synthesis

Synonym(s):

3-Methoxybenzaldehyde, m-Anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(CH3O)C6H4CHO
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352114
EC Index Number:
209-712-8

vapor pressure

1.3 hPa ( 73 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

225 °C

bp

230-233 °C/1013 hPa

transition temp

flash point 110 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H8O2/c1-10-8-4-2-3-7(5-8)6-9/h2-6H,1H3

InChI key

WMPDAIZRQDCGFH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.116 - 1.120
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service