Skip to Content
Merck
All Photos(1)

Key Documents

8.04812

Sigma-Aldrich

Methylenesuccinic acid

for synthesis

Synonym(s):

Methylenesuccinic acid, Itaconic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 26.00
500 G
CHF 36.80
50 KG
CHF 1’470.00

CHF 26.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 G
CHF 26.00
500 G
CHF 36.80
50 KG
CHF 1’470.00

About This Item

Linear Formula:
HOOCCH2C(=CH2)COOH
CAS Number:
Molecular Weight:
130.10
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-599-6
NACRES:
NA.22

CHF 26.00


Estimated to ship on29 May 2025


vapor pressure

0.0000115 Pa ( 20 °C)

Quality Level

Assay

≥99.0% (HPLC)

autoignition temp.

430 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

2 (20 °C, 10 g/L in H2O)

mp

163-168.4 °C

solubility

77.49 g/L

density

0.949 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CC(=C)C(=O)[O-].[H+].[H+]

InChI

1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h1-2H2,(H,6,7)(H,8,9)

InChI key

LVHBHZANLOWSRM-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 163 °C
Melting range (upper value): ≤ 167 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service