Skip to Content
Merck
All Photos(1)

Key Documents

8.03227

Sigma-Aldrich

1,4-Dichlorobutane

for synthesis

Synonym(s):

1,4-Dichlorobutane, Tetramethylene chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
CHF 45.40
1 L
CHF 123.00

CHF 45.40


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
250 ML
CHF 45.40
1 L
CHF 123.00

About This Item

Linear Formula:
Cl(CH2)4Cl
CAS Number:
Molecular Weight:
127.01
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-778-1
NACRES:
NA.22

CHF 45.40


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

220 °C

potency

3420 mg/kg LD50, oral (Rat)

expl. lim.

1.8-8.9 % (v/v)

pH

7.3 (20 °C in H2O)

bp

155 °C/1013 hPa

mp

-38 °C

transition temp

flash point 53 °C

density

1.14 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCCCCCl

InChI

1S/C4H8Cl2/c5-3-1-2-4-6/h1-4H2

InChI key

KJDRSWPQXHESDQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.136 - 1.140
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service