Skip to Content
Merck
All Photos(1)

Key Documents

8.03202

Sigma-Aldrich

Epoxystyrene

for synthesis

Synonym(s):

Epoxystyrene, Styrene oxide, Phenyloxirane, Phenylethylene oxide, Styrene epoxide

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
CHF 27.90
250 ML
CHF 52.30

CHF 27.90


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 ML
CHF 27.90
250 ML
CHF 52.30

About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-476-7
NACRES:
NA.22

CHF 27.90


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

498 °C

expl. lim.

1.1-22 % (v/v)

bp

193-195 °C/1013 hPa

mp

-37 °C

transition temp

flash point 74 °C

solubility

3 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(C1)c2ccccc2

InChI

1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2

InChI key

AWMVMTVKBNGEAK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.051 - 1.054
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service