Skip to Content
Merck
All Photos(1)

Key Documents

8.01079

Sigma-Aldrich

Aluminium triisopropylate

for synthesis

Synonym(s):

Aluminium triisopropylate, Aluminium isopropoxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
CHF 55.50
1 KG
CHF 110.00
20 KG
CHF 738.00
50 KG
CHF 1’650.00

CHF 55.50


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
250 G
CHF 55.50
1 KG
CHF 110.00
20 KG
CHF 738.00
50 KG
CHF 1’650.00

About This Item

Linear Formula:
Al[OCH(CH3)2]3
CAS Number:
Molecular Weight:
204.24
MDL number:
UNSPSC Code:
12352103
EC Index Number:
209-090-8
NACRES:
NA.22

CHF 55.50


Only 0 left in stock

Request a Bulk Order

vapor pressure

0.13 hPa ( 21 °C)

Quality Level

form

powder

potency

11300 mg/kg LD50, oral (Rat)

expl. lim.

2 % (v/v)

reaction suitability

reagent type: reductant

bp

135 °C/13 hPa

mp

130-134 °C

transition temp

flash point 16 °C

density

1.035 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Al](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/3C3H7O.Al/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

SMZOGRDCAXLAAR-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 98.0 %
Melting range (lower value): ≥ 127 °C
Melting range (upper value): ≤ 135 °C
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service