Skip to Content
Merck
All Photos(1)

Key Documents

8.00610

Sigma-Aldrich

Hydroxypropyl methacrylate

(stabilised)

Synonym(s):

Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
CHF 41.30
1 L
CHF 118.00

CHF 41.30


Estimated to ship on08 May 2025



Select a Size

Change View
250 ML
CHF 41.30
1 L
CHF 118.00

About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH(OH)CH3
CAS Number:
Molecular Weight:
144.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
248-666-3

CHF 41.30


Estimated to ship on08 May 2025


vapor pressure

0.1 hPa ( 20 °C)

Quality Level

autoignition temp.

355 °C

potency

>4000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

6 (20 °C, 50 g/L in H2O)

bp

240 °C/1013 hPa

mp

-58 °C

transition temp

flash point 101 °C

solubility

107 g/L

density

1.029 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(CO)C)C(=O)C(=C)C

InChI

1S/C7H12O3/c1-5(2)7(9)10-6(3)4-8/h6,8H,1,4H2,2-3H3

InChI key

ZMARGGQEAJXRFP-UHFFFAOYSA-N

Analysis Note

Assay isomers (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.027 - 1.030
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service