Skip to Content
Merck
All Photos(2)

Key Documents

8.00218

Sigma-Aldrich

(-)-Quinic acid

for resolution of racemates for synthesis

Synonym(s):

(-)-Quinic acid, 1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
CHF 116.00

CHF 116.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
CHF 116.00

About This Item

Empirical Formula (Hill Notation):
C7H12O6
CAS Number:
Molecular Weight:
192.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-072-8
NACRES:
NA.22

CHF 116.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

pH

2.0 (20 °C, 10 g/L in H2O)

mp

162.5 °C

solubility

290 g/L (experimental)

density

1.64 g/cm3 at 20 °C

bulk density

670‑750 kg/m3

storage temp.

2-30°C

SMILES string

OC1(CC(C(C(C1)O)O)O)C(=O)O

InChI

1S/C7H12O6/c8-3-1-7(13,6(11)12)2-4(9)5(3)10/h3-5,8-10,13H,1-2H2,(H,11,12)

InChI key

AAWZDTNXLSGCEK-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 160 °C
Melting range (upper value): ≤ 165 °C
Spec. rotation [α²0/D (c=10 in water): -44.0 - -42.0 °
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service