Skip to Content
Merck
All Photos(1)

Key Documents

8.00105

Sigma-Aldrich

2,5-Hexanedione

for synthesis

Synonym(s):

2,5-Hexanedione, Acetonylacetone

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 95.30
250 ML
CHF 159.00

CHF 95.30


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
CHF 95.30
250 ML
CHF 159.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-738-3
NACRES:
NA.22

CHF 95.30


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

0.6 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

400 °C

potency

1600 mg/kg LD50, oral (Rat)

expl. lim.

1.5 % (v/v)

pH

6.1 (20 °C, 10 g/L in H2O)

bp

85-87 °C/26.6 hPa

mp

-5.4 °C

transition temp

flash point 79 °C

density

0.974 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CCC(=O)C)C

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.971 - 0.973
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service