Skip to Content
Merck
All Photos(1)

Key Documents

73293BC

Millipore

mEHEC® Media Level Assembly

BioControl®, For use with GDS

Sign Into View Organizational & Contract Pricing

Select a Size

73293-SIAL
CHF 575.00

CHF 575.00


Estimated to ship on10 May 2025



Select a Size

Change View
73293-SIAL
CHF 575.00

About This Item

UNSPSC Code:
41121521
NACRES:
NA.74

CHF 575.00


Estimated to ship on10 May 2025


storage condition

room temperature

Quality Level

compatibility

BioControl®, For use with GDS

Components

One media leveler

Testing Method

PCR

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
mEHEC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service