Skip to Content
Merck
All Photos(1)

Key Documents

71131

Sigma-Aldrich

pETcoco-1 System - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 740.00

CHF 740.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
CHF 740.00

About This Item

UNSPSC Code:
41106602
NACRES:
NA.54

CHF 740.00


Estimated to ship on05 May 2025


Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

shipped in

dry ice

storage temp.

−70°C

General description

The pETcoco System combines the advantages of T7 promoter-driven proteinexpression with the ability to control plasmid copy number (Sektas 2003). Inthe presence of 0.2% glucose, pETcoco recombinant plasmids are maintained atone copy per cell. In this single-copy state, pETcoco clones are extremely stable becausebackground expression and consequent gene rearrangements or deletions are virtuallyabsent. For plasmid preparation, the copy number can be amplified to approximately 40copies per cell independently of target gene expression by addition of L-arabinose to theculture medium. Both target gene expression and plasmid plus insert amplificationare induced upon addition of IPTG. Both pETcoco-1 and pETcoco-2 vectors featurean N-terminal His•Tag and internal S•Tag sequences, both of which are cleavable by enterokinase, and an optional C-terminal HSV•Tag sequence. NovaF- Competent Cells for cloning target genes and Tuner(DE3) Competent Cells for expression are supplied with the pETcoco-1 and pETcoco-2 Systems.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled.

Features and Benefits

  • T7 promoter-driven protein expression
  • Offers the ability to control plasmid copy number
  • Works with toxic proteins
  • N-terminal HIS TAG® sequence
  • Internal S-Tag sequence
  • Optional C-termainal HSV-Tag sequence

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service