Skip to Content
Merck
All Photos(1)

Key Documents

70923

Millipore

BugBuster® (primary amine-free) Extraction Reagent

Synonym(s):

Protein Extraction Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 118.00
500 ML
CHF 474.00

CHF 118.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 ML
CHF 118.00
500 ML
CHF 474.00

About This Item

UNSPSC Code:
41106511
NACRES:
NA.77

CHF 118.00


Estimated to ship on25 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

10-30°C

General description

BugBuster (primary amine-free) Protein Extraction Reagent is a special formulation of BugBuster designed for applications where primary amines would interfere if present in the protein extract, such as protein immobilization or cross-linking. The PIPPS buffer used in BugBuster (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins.

Features and Benefits

Primary Amine-Free Formulation: BugBuster® (primary amine-free) Extraction Reagent is specially designed for applications where primary amines would interfere with protein extract. This means that academic researchers can purify metal-dependent proteins or proteins to be used for immobilization or crosslinking without worrying about the presence of primary amines, which could cause interference.

Enhanced Extraction: The extraction process can be further enhanced by the addition of rLysozyme Solution and Benzonase Nuclease, which helps to efficiently extract and purify proteins from the sample.

Metal Ion Compatibility: The PIPPS buffer used in the primary amine-free formulation of BugBuster® has a similar buffer capacity and pH range as the original Tris-buffered BugBuster® but will not complex metal ions. This makes it an ideal choice for extraction of metal-dependent proteins

Warning

Toxicity: Standard Handling (A)

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Citation Spotlight: BugBuster® Protein Extraction Reagent for Efficient Protein Extraction from Bacterial Pathogens

Cell lysis and nucleic acid removal for Gram-negative and Gram-positive bacteria using the BugBuster Plus Lysonase™ Kit

The combination of BugBuster® and Lysonase™ reagents greatly enhances the release of soluble proteins from Gram-positive bacteria.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service