Skip to Content
Merck
All Photos(1)

Key Documents

70099-M

Sigma-Aldrich

T4 DNA Polymerase, LIC-qualified - Novagen

Novagen′s T4 DNA Polymerase has been qualified for ligation-independent cloning using a unique assay to verify its performance.

Synonym(s):

T4 Polymerase

Sign Into View Organizational & Contract Pricing

Select a Size

250 U
CHF 768.00

CHF 768.00


Estimated to ship on31 March 2025



Select a Size

Change View
250 U
CHF 768.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

CHF 768.00


Estimated to ship on31 March 2025


Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

molecular cloning: suitable

storage temp.

−20°C

General description

T4 DNA polymerase is a potent enzymatic tool used in the process of ligation-independent cloning (LIC), a technique that allows to insert DNA fragments into plasmid vectors without the need for traditional restriction enzyme digestion and ligation steps. The 3′ exonuclease activity of T4 DNA polymerase is used in the LIC process to create single-stranded overhangs on the DNA fragment, which can then anneal with complementary overhangs on the vector, allowing the fragment to be inserted. In the LIC process, T4 DNA polymerase, in conjunction with complementary primers designed to lack one of dATP, dTTP, dCTP, or dGTP for a sequence of 12–15 bp, is used to generate single-stranded complementary overhangs of a fixed length. This annealing technique can then be followed by transformation directly into E. coli, which repairs the nicked DNA backbone, resulting in the successful insertion of the DNA fragment into the vector[1]

Application

T4 DNA Polymerase, LIC-qualified – Novagen has been used in the ligation-independent cloning (LIC) reaction to generate single-stranded 5′ termini overhangs on the DNA insert and destination plasmid vector.[2][3][4]

Components

•250 UT4 DNA Polymerase, LIC-qualified

•500 µl10X T4DNA Polymerase Buffer

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service