Skip to Content
Merck
All Photos(1)

Key Documents

681674

Sigma-Aldrich

Wnt Pathway Inhibitor XI, CCT036477

The Wnt Pathway Inhibitor XI, CCT036477, also referenced under CAS 305372-78-5, controls the biological activity of Wnt Pathway. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Wnt Pathway Inhibitor XI, CCT036477, CCT036477, CRT Inhibitor VI, β-Catenin/Tcf Inhibitor VI

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 139.00

CHF 139.00


Estimated to ship on07 April 2025



Select a Size

Change View
5 MG
CHF 139.00

About This Item

Empirical Formula (Hill Notation):
C21H18ClN3
CAS Number:
Molecular Weight:
347.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 139.00


Estimated to ship on07 April 2025


Quality Level

Assay

≥95% (1H-NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(Nc4ncccc4)c2c3c([nH]c2C)cccc3

InChI

1S/C21H18ClN3/c1-14-20(17-6-2-3-7-18(17)24-14)21(15-9-11-16(22)12-10-15)25-19-8-4-5-13-23-19/h2-13,21,24H,1H3,(H,23,25)

InChI key

IFOHRIXRQPOHQZ-UHFFFAOYSA-N

General description

A cell-permeable indole derivative that acts as an inhibitor of Wnt signaling and exhibits an inhibitory effect in a reporter cell line (IC50 = 4.6 µM) and demonstrates growth inhibition in multiple cancer cell lines, not by altering β-catenin accumulation but by blocking transcription at the β-catenin level. Displays reduced expression of Wnt target genes in myc and survivin-luciferase reporter assays, and exhibits in vivo activity by blocking the development of zebrafish at 20 µM and Xenopus embryos at 75 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following recontitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ewan, K., et al. 2010. Cancer Res.70, 5963.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service