Skip to Content
Merck
All Photos(1)

Key Documents

6680

Millipore

Uridine

Synonym(s):

Uridine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
Inquire
100 G
Inquire


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
Inquire
100 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51


Estimated to ship on29 March 2025


Request a Bulk Order

Assay

≥97% (HPLC)
≥99% (UV)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1% ash

color

white to off-white

solubility

water: soluble

cation traces

heavy metals: <10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[nH]1[c]([n](cc[c]1=O)C2OC(C(C2O)O)CO)=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)

InChI key

DRTQHJPVMGBUCF-UHFFFAOYSA-N

Application


  • Modified Nucleotides and RNA Structure Prediction.: This research explores the impact of modified nucleotides, including uridine analogs, on RNA structure prediction. The study advances our understanding of RNA biology and the role of uridine modifications in gene expression and regulation, highlighting the importance of uridine in biochemical research (Varenyk and Lorenz, 2024).

  • CAD-Related Disorder (EIEE-50) in an Infant With Cortical Visual Impairment.: This article discusses the application of uridine in the context of CAD-related disorders. The study suggests that uridine supplementation may have therapeutic benefits for infants with cortical visual impairment and related metabolic disorders (Thurman et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service