Skip to Content
Merck
All Photos(1)

Documents

662065

Sigma-Aldrich

Ubiquitin, Methylated, Human, Recombinant

Synonym(s):

Me-Ub

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

solubility

aqueous buffer: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human methylated ubiquitin. A derivative of ubiquitin in which lysine residues are reductively methylated. Methylated ubiquitin can ligate to protein substrates, but does not form polyubiquitin chains. Protein breakdown rates obtained with methylated ubiquitin are slower than those obtained with unmethylated ubiquitin. Useful for reducing the polyubiquitin chain length and for determining rates of ubiquitin conjugation.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hershko, A. and Heller, H. 1991. J. Biol. Chem.266, 16376.
Hershko, A. and Heller, H. 1985. Biochem. Biophys. Res. Commun.128, 1079.
Jentoft, N. and Dearborn, D.G. 1983. Methods. Enz.91, 570.
Jentoft, N. and Dearborn, D.G. 1980. Anal. Biochem.106, 186.
Jentoft, N. and Dearborn, D.G. 1979. J. Biol. Chem.254, 4359.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service