Skip to Content
Merck
All Photos(1)

Key Documents

658551

Sigma-Aldrich

AG 1296

A cell-permeable, reversible, and ATP-competitive, inhibitor of PDGF receptor kinase and blocks signaling of human PDGF α-receptors and β-receptors.

Synonym(s):

AG 1296, 6,7-Dimethoxy-3-phenylquinoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
acetonitrile: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C16H14N2O2/c1-19-15-8-12-13(9-16(15)20-2)18-14(10-17-12)11-6-4-3-5-7-11/h3-10H,1-2H3

InChI key

QNOXYUNHIGOWNY-UHFFFAOYSA-N

General description

Similar to but more potent than AG 1295 (Cat. No. 658550). A cell-permeable, reversible, and ATP-competitive, inhibitor of PDGF receptor kinase and blocks signaling of human PDGF α-receptors (IC50 = 1.0 µM) and β-receptors (IC50 = 800 nM) as well as of the related stem cell factor receptor c-kit (80% inhibition at 5 µM). Affects neither the binding of PDGF-BB to the PDGF-β receptor nor the receptor dimerization. Has no effect on autophosphorylation of the vascular endothelial growth factor receptor KDR or on DNA synthesis induced by vascular endothelial growth factor in porcine aortic endothelial cells. Also reverses the transformed phenotype of sis-transfected NIH/3T3 cells but has no effect on src-transformed NIH 3T3 cells or on the activity of the kinase p60c-src (F257) immunoprecipitated from these cells. Potently inhibits autophosphorylation in Flt3/Tel-BaF3 and Flt3/Itd-BaF3 cells with an IC50 of 300 nM and ~1 µM, respectively.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDGFR kinase
Product competes with ATP.
Reversible: yes
Target IC50: 1.0 µM, 800 nM, against signaling of human PDGF α-receptors, human PDGF β-receptors, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Tse, KF., et al. 2002. Leukemia16, 2027.
Tse, KF., et al. 2001. Leukemia15, 1001.
Strutz, F., et al. 2001. Kidney Int.59, 579.
Kovalenko, M., et al. 1997. Biochemistry 36, 6260.
Kovalenko, M., et al. 1994. Cancer Res. 54, 6106.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sen Jiang et al.
Biomedicines, 10(10) (2022-10-28)
A major symptom of diabetes mellitus (DM) is unfit hyperglycemia, which leads to impaired wound healing. It has been reported that the migration of fibroblasts can be suppressed under high glucose (HG) conditions. In our previous study, we introduced a
Mariana S Dias et al.
Gene therapy, 26(12), 479-490 (2019-09-29)
Adeno-associated virus vectors (rAAV) are currently the most common vehicle used in clinical trials of retinal gene therapy, usually delivered through subretinal injections to target cells of the outer retina. However, targeting the inner retina requires intravitreal injections, a simple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service