Skip to Content
Merck
All Photos(1)

Key Documents

654271

Sigma-Aldrich

MetAP2 Inhibitor, A832234 - Calbiochem

Synonym(s):

Methionine aminopeptidase-2 Inhibitor, 2-[({2-[(1Z)-3-(Diethylamino)-1-propenyl]-4-fluorophenyl}-sulfonyl)amino]-5,6,7,8-tetrahydro-1-naphthalenecarboxylic Acid, Methionyl aminopeptidase 2 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥97% (HPLC)

form

solid

potency

11 nM IC50

color

light beige

solubility

DMSO: 5 mg/mL, pale yellow

General description

A cell-permeable anthranilic acid sulfonamide compound that acts as a potent and reversible inhibitor of methionine aminopetidase-2 (MetAP2, IC50 = 11 nM). Acts by directly targeting the active site of MetAP2 without any significant binding to human serum albumin. Also shown to inhibit cellular MetAP2 activity (EC50 = 5.3 nM) and block the proliferation of HT1080 fibrosarcoma cells (EC50 = 6 nM).
The MetAP2 Inhibitor, A832234 controls the biological activity of MetAP2.

Biochem/physiol Actions

MetAP2

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George S Sheppard et al.
Journal of medicinal chemistry, 49(13), 3832-3849 (2006-06-23)
Methionine aminopeptidase-2 (MetAP2) is a novel target for cancer therapy. As part of an effort to discover orally active reversible inhibitors of MetAP2, a series of anthranilic acid sulfonamides with micromolar affinities for human MetAP2 were identified using affinity selection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service