Skip to Content
Merck
All Photos(1)

Key Documents

616459

Sigma-Aldrich

TGF-β RI Kinase Inhibitor VIII

The TGF-β RI Kinase Inhibitor VIII, also referenced under CAS 356559-20-1, controls the biological activity of TGF-β RI Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

TGF-β RI Kinase Inhibitor VIII, ALK5 Inhibitor VIII, SB-525334, 6-(2-tert-Butyl-5-(6-methyl-pyridin-2-yl)-1H-imidazol-4-yl)-quinoxaline, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N5
CAS Number:
Molecular Weight:
343.42
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C21H21N5/c1-13-6-5-7-16(24-13)19-18(25-20(26-19)21(2,3)4)14-8-9-15-17(12-14)23-11-10-22-15/h5-12H,1-4H3,(H,25,26)

InChI key

DKPQHFZUICCZHF-UHFFFAOYSA-N

General description

A cell-permeable quinoxaline compound that acts as a potent, highly selective, ATP-competitive inhibitor against type I TGF-β receptor TβR-I/ALK5 (IC50 = 14.3 nM), while exhibiting 4-fold less potency toward ALK4 (IC50 = 58.5 nM) and little or no activity against ALK2/3/6 and a panel of 26 other commonly studied kinases (≤29% inhibition at 10 µM). Effectively blocks (by >95%) TGF-β1-induced Smad2/3 activation (2 µM in ELT-3 cultures) and nuclear localization (1 µM in RPTE cultures), as well as TGF-β1-dependent p3TP- and ARE-luciferase transcription (0.1 µM in HaCat cultures). SB-525334 is orally available and is shown to attenuate Bleomycin- and Puromycin-induced tissue fibrosis in mice and rats in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20&deg:C). Stock solution is stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Higashiyama, H., et al. 2007. Exp. Mol. Pathol.83, 39.
Grygielko, E.T., et al. 2005. J. Pharmacol. Exp. Ther.313, 943.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service