Skip to Content
Merck
All Photos(1)

Key Documents

614852

Sigma-Aldrich

Topoisomerase IIα Inhibitor, TSC24

The Topoisomerase IIα Inhibitor, TSC24 controls the biological activity of Topoisomerase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Topoisomerase IIα Inhibitor, TSC24, (E)-N,N-dimethyl-2-(quinolin-2-ylmethylene)hydrazinecarbothioamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 155.00

CHF 155.00


Estimated to ship on01 June 2025



Select a Size

Change View
10 MG
CHF 155.00

About This Item

Empirical Formula (Hill Notation):
C13H14N4S
Molecular Weight:
258.34
UNSPSC Code:
12352200
NACRES:
NA.54

CHF 155.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

DMSO: 15 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, TSC (thiosemicarbazone) derivative that acts as a selective and ATP competitive Topoisomerase IIα inhibitor and Fe chelator. Independent of its Fe chelating ability, this compound is shown to block Topo II&alpha-catalyzed ATP hydrolysis by competing with ATP for binding to the ATPase domain of Topo IIα (Kd = 18 µM). It also displays M phase cell-cycle arrest at 1 µM in nocodazole-treated HT-29 cells. This compound demonstrates anti-proliferative activity, without distinct or obvious selectivity, towards HeLa, HL-60, SGC7901, and HT-29 cultures (IC50 = 90 nM, 50 nM, 30 nM, and 20 nM, respectively), and a broader panel of tumor cell lines with IC50 values in the nanomolar range.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huang, H., et al. 2010. J. Med. Chem.53, 3048.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service