Skip to Content
Merck
All Photos(1)

Key Documents

574714

Sigma-Aldrich

BAY 61-3606

≥97% (HPLC), solid, Syk inhibitor, Calbiochem®

Synonym(s):

Syk Inhibitor IV, BAY 61-3606, 2-(7-(3,4-Dimethoxyphenyl)-imidazo[1,2-c]pyrimidin-5-ylamino)-nicotinamide, HCl, BAY 61-3606

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
CHF 333.00

CHF 333.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
2 MG
CHF 333.00

About This Item

Empirical Formula (Hill Notation):
C20H18N6O3 · xHCl · yH2O
CAS Number:
Molecular Weight:
390.40 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 333.00


Estimated to ship on23 May 2025


Request a Bulk Order

Product Name

Syk Inhibitor IV, BAY 61-3606, The Syk Inhibitor IV, BAY 61-3606, also referenced under CAS 732938-37-8, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

10 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21c(ncc2)cc(nc1Nc4ncccc4C(=O)N)c3cc(c(cc3)OC)OC

InChI

1S/C20H18N6O3/c1-28-15-6-5-12(10-16(15)29-2)14-11-17-22-8-9-26(17)20(24-14)25-19-13(18(21)27)4-3-7-23-19/h3-11H,1-2H3,(H2,21,27)(H,23,24,25)

InChI key

JWQOJVOKBAAAAR-UHFFFAOYSA-N

General description

A cell-permeable imidazopyrimidine compound that acts a potent, ATP-competitive, reversible, and highly selective inhibitor of Syk tyrosine kinase activity (IC50 = 10 nM) with no inhibitory effect against Btk, Fyn, Itk, Lyn, and Src even at concentrations as high as 4.7 µM. Shown to inhibit Syk-mediated cellular functions in vitro and exhibit good oral bioavailability and in vivo effacacy in the treatment of various allgery and asthma conditions in rat models.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Syk
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yamamoto, N., et al. 2003. J. Pharm. Exp. Ther.306, 1174.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haruka Miki et al.
Journal of immunology (Baltimore, Md. : 1950), 204(4), 753-762 (2020-01-05)
House dust mite (HDM) allergens are leading causes of allergic asthma characterized by Th2 responses. The lung-resident CD11b+ dendritic cells (DCs) play a key role in Th2 cell development in HDM-induced allergic asthma. However, the regulatory mechanism of HDM-induced CD11b+
Ilaria Frasson et al.
Journal of molecular cell biology, 16(3) (2024-02-02)
The high mutation rate of SARS-CoV-2 leads to the emergence of multiple variants, some of which are resistant to vaccines and drugs targeting viral elements. Targeting host dependency factors, e.g. cellular proteins required for viral replication, would help prevent the
Sinu P John et al.
Cell reports, 41(1), 111441-111441 (2022-10-01)
Biologically active small molecules can impart modulatory effects, in some cases providing extended long-term memory. In a screen of biologically active small molecules for regulators of tumor necrosis factor (TNF) induction, we identify several compounds with the ability to induce
Ei'ichi Iizasa et al.
Nature communications, 12(1), 2299-2299 (2021-04-18)
Mycobacterial cell-wall glycolipids elicit an anti-mycobacterial immune response via FcRγ-associated C-type lectin receptors, including Mincle, and caspase-recruitment domain family member 9 (CARD9). Additionally, mycobacteria harbor immuno-evasive cell-wall lipids associated with virulence and latency; however, a mechanism of action is unclear.
Rohan P Choraghe et al.
Journal of cell science, 133(5) (2020-01-23)
Dendritic cell-associated C-type lectin 1 (Dectin-1, also known as CLEC7A) is an innate immune pattern recognition receptor that recognizes β-glucan on the Candida albicans cell wall. Recognition of β-glucan by immune cells leads to phagocytosis, oxidative burst, cytokine and chemokine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service