Skip to Content
Merck
All Photos(1)

Key Documents

573125

Sigma-Aldrich

STAT3 Inhibitor IX, Cpd188

The STAT3 Inhibitor IX, Cpd188, also referenced under CAS 823828-18-8, controls the biological activity of STAT3.

Synonym(s):

STAT3 Inhibitor IX, Cpd188, 4-((3-(Carboxymethylsulfanyl)-4-hydroxy-1-naphthyl)sulfamoyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO7S2
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2c3c(c(c(c2)SCC(=O)O)O)cccc3)c1ccc(cc1)C(=O)O

InChI

1S/C19H15NO7S2/c21-17(22)10-28-16-9-15(13-3-1-2-4-14(13)18(16)23)20-29(26,27)12-7-5-11(6-8-12)19(24)25/h1-9,20,23H,10H2,(H,21,22)(H,24,25)

InChI key

CIERPRDYPKVDJO-UHFFFAOYSA-N

General description

A cell-permeable naphthol compound that inhibits IL-6-stimulated STAT3 Tyr705 phosphorylation and nuclear translocation in HepG2 cells (IC50 = 73 and 39 µM, respectively) by targeting STAT3 SH2 domain peptide-binding pocket (IC50 = 20 µM against EGFR-derived pY-peptide binding), while exhibiting little effect against IFN-γ-stimulated HepG2 cellular STAT1 phosphorylation even at concentrations as high as 300 µM. Shown to preferentially induce apoptosis in cultures expressing constitutively active STAT3 (EC50 = 0.73, 3.96, and 7.01 µM in MDA-MB-468, MDA-MB-231, and MDA-MB-435 cultures, respectively), while being less effective against cells without constitutively active STAT3 (EC50 = 15.5 and 17.19 µM in MDA-MB-453 and MCF7 cultures, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xu, X., et al. 2009. PLoS One4, e4783.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service