Skip to Content
Merck
All Photos(1)

Documents

566324

Sigma-Aldrich

SIRT2 Inhibitor, AGK2

The SIRT2 Inhibitor, AGK2, also referenced under CAS 304896-28-4, controls the biological activity of SIRT2. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT2 Inhibitor, AGK2, 2-Cyano-3-(5-(2,5-dichlorophenyl)-2-furanyl)-N-5-quinolinyl-2-propenamide, Sirtuin 2 Inhibitor, SIRT2 Inhibitor I, SIRT1 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H13Cl2N3O2
CAS Number:
Molecular Weight:
434.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 2.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H13Cl2N3O2/c24-15-6-8-19(25)18(12-15)22-9-7-16(30-22)11-14(13-26)23(29)28-21-5-1-4-20-17(21)3-2-10-27-20/h1-12H,(H,28,29)/b14-11+

InChI key

SVENPFFEMUOOGK-SDNWHVSQSA-N

General description

A cell-permeable quinoline compound that targets the nicotinamide-binding site and acts as a reversible sirtuin 2- (SIRT2) selective inhibitor (IC50 = 3.5 µM) with little activity against SIRT1/3 (IC50 >50 µM). Prevents cellular α-tubulin deacetylation in HeLa cells in a dose-dependent manner and with little cytotoxic effect. Shown to rescue α-Synuclein-mediated toxicity both in primary rat embryo midbrain cultures in vitro and in a Drosophila PD model in vivo. The inactive isomer AGK7 (Cat. No. 566326) can be used as a negative control.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required for complete solubilization.

Other Notes

Outerio, T.F., et al. 2007. Science317, 516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Carlos Gómora-García et al.
Cells, 12(3) (2023-02-12)
Mitochondrial activity and quality control are essential for neuronal homeostasis as neurons rely on glucose oxidative metabolism. The ketone body, D-β-hydroxybutyrate (D-BHB), is metabolized to acetyl-CoA in brain mitochondria and used as an energy fuel alternative to glucose. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service