565840
SHIP2 Inhibitor, AS1938909
The SHIP2 Inhibitor, AS1938909 controls the biological activity of SHIP2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
SHIP2 Inhibitor, AS1938909, 3-(2,4-Dichlorobenzyl)oxy]-N-(2,6-difluorobenzyl)thiophene-2-carboxamide, SH2 domain-containing inositol 5ʹ-phosphatase 2 (SHIP2) Inhibitor, AS1938909
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 50 mg/mL
shipped in
wet ice
storage temp.
2-8°C
General description
Packaging
Warning
Other Notes
Suwa, A., et al. 2009. Br. J. Pharmacol.158, 879.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service