Skip to Content
Merck
All Photos(1)

Key Documents

558300

Millipore

Aquatic Block Reagent, Salmon Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
CHF 225.00

CHF 225.00


Estimated to ship on23 May 2025



Select a Size

Change View
500 ML
CHF 225.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.77

CHF 225.00


Estimated to ship on23 May 2025


form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

A ready-to-use blocking reagent containing non-mammalian proteins that do not interact with mammalian antibodies.
A ready-to-use blocking reagent for Western blotting and immunoassays. Contains salmon plasma proteins that do not interact with mammalian antibodies, thus offering a reduced background.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.09% NaN₃, pH 7.4.

Legal Information

AQUATIC BLOCK is a trademark of Arlington Scientific, Inc.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service