Skip to Content
Merck
All Photos(1)

Key Documents

539636

Sigma-Aldrich

PKCθ Pseudosubstrate Inhibitor, Myristoylated

The PKCθ Pseudosubstrate Inhibitor, Myristoylated controls the biological activity of PKCθ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKCθ Pseudosubstrate Inhibitor, Myristoylated, Myr-LHQRRGAIKQAKVHHVKC-NH₂, Protein Kinase C θ Pseudosubstrate Inhibitor, Myristoylated, Myr-LHQRRGAIKQAKVHHVKC-NH₂, Protein Kinase Cθ Pseudosubstrate Inhibitor, Myristoylated

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
CHF 276.00

CHF 276.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 μG
CHF 276.00

About This Item

Empirical Formula (Hill Notation):
C105H182N35O22S
Molecular Weight:
2318.85
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 276.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible, substrate competitive, myristoylated form of the inhibitor of protein kinase Cθ isozyme pseudosubstrate inhibitor (Cat. No. 539634).
Cell-permeable myristoylated protein kinase Cq pseudosubstrate sequence peptide (Cat. No. 539634). Useful for studies of PKCθ function in intact cells. Sold on the basis of peptide content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
protein kinase Cθ
Product competes with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-Leu-His-Gln-Arg-Arg-Gly-Ala-Ile-Lys-Gln-Ala-Lys-Val-His-His-Val-Lys-Cys-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 6 months at -20°C.

Other Notes

Osada, S., et al. 1992. Mol. Cell Biol.12, 3930.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service